Learn More
OHAUS™ Uni Block
Shop All Ohaus Products
missing translation for 'orderingAttributeHoverText'
For Use With (Equipment):
12 mm Test Tubes
12 mm Vials
15 mm Vials
16 mm Test Tubes
17 mm Vials
20 mm Test Tubes
21 mm Vials
25 mm Test Tubes
28 mm Vials
missing translation for 'unitSize'
Each
Description
Holds test tubes or vials
Specifications
Specifications
| For Use With (Equipment) | 12 mm Test Tubes |
| Type | Uni Block |
| Material | Metal |