Learn More
Haldenwanger™ Unglazed Porcelain Pestle
Click to view available options
Length (Metric):
115mm
125mm
135mm
150mm
175mm
Unit Size:
Each
Description
- Available in a variety of sizes
- Grinding surfaces are unglazed
- Complies with DIN VDE 0335, type C 100
Specifications
Specifications
| Finish | Unglazed |
| Product Type | Pestle |
| Material | Porcelain |
| Length (Metric) | 135mm |