missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UNC5B Polyclonal Antibody
GREENER_CHOICE

Product Code. 15527532
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15527532 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15527532 Supplier Invitrogen™ Supplier No. PA547038

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) UNC5H3 and recombinant rat UNC5H1 is observed, and less than 1% cross-reactivity with rhUNC5H4 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

Receptor for netrin required for axon guidance. Mediates axon repulsion of neuronal growth cones in the developing nervous system upon ligand binding. Axon repulsion in growth cones may be caused by its association with DCC that may trigger signaling for repulsion. It also acts as a dependence receptor required for apoptosis induction when not associated with netrin ligand. Mediates apoptosis by activating DAPK1. In the absence of NTN1, activates DAPK1 by reducing its autoinhibitory phosphorylation at Ser-308 thereby increasing its catalytic activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UNC5B
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene UNC5B
Gene Accession No. O08722
Gene Alias 6330415E02Rik; A630020F16; C. elegans; D10Bwg0792e; Netrin receptor UNC5B; p53RDL1; p53-regulated receptor for death and life protein 1; Protein unc-5 homolog 2; protein unc-5 homolog B; transmembrane receptor Unc5H2; unc5 homolog 2; unc-5 homolog 2; unc-5 homolog B; unc-5 homolog B (C. elegans); unc-5 netrin receptor B; Unc5b; UNC5H2; UNQ1883/PRO4326
Gene Symbols UNC5B
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant rat UNC5H2 Gly27-Asp373.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60630
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.