missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UNC2025 hydrochloride, MedChemExpress
Description
UNC2025 hydrochloride is a potent, ATP-competitive, and highly orally active Mer/Flt3 inhibitor with IC50 values of 0.74 nM and 0.8 nM, respectively. UNC2025 hydrochloride is >45-fold selectivity for MERTK relative to Axl (IC50= 122 nM
Specifications
Specifications
| Chemical Name or Material | UNC2025 hydrochloride |
| CAS | 2070015-17-5 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C28H41ClN6O |
| Quantity | 10 mg |
| Solubility Information | H2O : 55 mg/mL (107.19 mM; Need ultrasonic) ∣DMSO : 10 mg/mL (19.49 mM; Need ultrasonic) |
| SMILES | CN1CCN(CC2=CC=C(C3=CN([C@@H]4CC[C@@H](O)CC4)C5=NC(NCCCC)=NC=C53)C=C2)CC1.[H]Cl |
| Molecular Weight (g/mol) | 513.12 |
| Formula Weight | 513.12 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?