missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UNC1079, MedChemExpress
Description
UNC1079 is the piperidine analog of UNC1021, as a structurally similar but significantly less potent inhibitor for use as a negative control in cellular studies.
Specifications
Specifications
| Chemical Name or Material | UNC1079 |
| CAS | 1418741-86-2 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C28H42N4O2 |
| Quantity | 100 mg |
| Solubility Information | DMSO : 2.22 mg/mL (4.76 mM; ultrasonic and warming and heat to 60°C) |
| SMILES | O=C(N1CCC(N2CCCCC2)CC1)C3=CC=C(C(N4CCC(N5CCCCC5)CC4)=O)C=C3 |
| Molecular Weight (g/mol) | 466.66 |
| Formula Weight | 466.66 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?