missing translation for 'onlineSavingsMsg'
Learn More

UNC 669, MedChemExpress

Product Code. 16476956
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16476956

Brand: Medchem Express HY15839

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress UNC 669, a ligand for a methyl-lysine binding domain, is a potent L3MBTL1 (IC50=4.2 uM) and L3MBTL3 (3.1 uM) inhibitor.

UNC 669, a ligand for a methyl-lysine binding domain, is a potent L3MBTL1 (IC50=4.2 uM) and L3MBTL3 (3.1 uM) inhibitor.

Specifications

Chemical Name or Material UNC 669
CAS 1314241-44-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C15H20BrN3O
Quantity 100 mg
Solubility Information DMSO : 50 mg/mL (147.82 mM; Need ultrasonic)
SMILES O=C(C1=CC(Br)=CN=C1)N2CCC(N3CCCC3)CC2
Molecular Weight (g/mol) 338.24
Formula Weight 338.24
Percent Purity 99.88%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.