Learn More
Emballator UN Approved Cans
Product Code. p-7083473
Encompass
Click to view available options
Capacity (Metric):
5 L
10 L
25 L
Height (Metric):
296 mm
306 mm
310 mm
467 mm
Unit Size:
Each
Product Code. 11394405
Brand: Emballator 48H0517410300
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Made of HDPE. Emballator UN Approved Cans approved by NET for storage and transport of dangerous goods.
  • Material: HDPE, approved for food contact
  • Color: Nature
  • UN Approved tamper evident closure
  • Approved by NET for storage and transport of dangerous goods
Specifications
Product Type UN Approved Can
Capacity (Metric) 5 L
Color Natural
Material HDPE
Length (Metric) 190 mm
Width (Metric) 130 mm
Height (Metric) 296 mm
Diameter (Metric) Outer 40 mm
Weight (Metric) 175 g
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado