Learn More
Thermo Scientific Ultralow Temp. Freezer -86C 549L - 400 Boxes 5 Inner Door
Brand:  Thermo Scientific TDE40086FV5I-U
6650.00 EUR valid until 2025-12-24
BEST PRICE on Promo! Use promo code "28321" to get your promotional price.
Ultralow Temp. Freezer -86C 549L - 400 Boxes 5 Inner Door
Product Code. 17945466
  • ONLINE EXCLUSIVE PRICE
    13570.00 EUR
    13300.00 EUR / Each
    Save 270.00 EUR
    1% Off