Learn More
Merck Ultrafiltration Membranes - Amicon™ XM Polyacrylonitrile/PVC Copolymer Filters
High-recovery membranes for stirred cells
Brand: Merck 14342
784.04 EUR valid until 2024-12-31
Use promo code "23269" to get your promotional price.
Additional Details : Weight : 0.10000kg
Description
- Regenerated cellulose membranes
- For concentration of hydrophobic samples or to desalt extremely dilute solutions
- Hydrophilic, tight microstructure assures high retention with low adsorption of protein, DNA, or other macromolecules
†43mm nominal diameter.
‡62mm nominal diameter.
Specifications
Polyacrylonitrile, PVC Co-polymer | |
High recovery membranes for stirred cells | |
Ultrafiltration Disc | |
7.6 cm |
300,000 NMWL | |
10 Pack | |
2.99 in. |