man
Test
Search
Order Status
Quick Order
Sign In
chevron-down
chevron-down
Learn More
Home
Products
Mixers
Blenders
Homogenizers and Immersion Blenders
13440597
Print
IKA™ ULTRA-TURRAX™ T 65 Digital Disperser
Brand:
IKA™ 0004234500
Additional Details :
Weight :
4.00000kg
Product Code.
13440597
9495.00 EUR
/ Each
Quantity
Estimated Shipment: 10-07-2024
Log in
Specifications
Product Type
Digital Disperser
Voltage
3 x 400 V
Hertz
50/60 Hz
Volume Range (Metric)
2 to 50 L
Height (Metric)
178 mm
Display
LED
Packaging Quantity
1 Ea.
Speed
1000 to 9500 rpm
Electrical Requirements
3 x 400 V, 50/60 Hz, 2600/2200 W
Wattage
2600/2200 W
Depth (Metric)
400 mm
Width (Metric)
300 mm
Model
T 65 ULTRA-TURRAX™
Viscosity
5000 mPas