Learn More
Sampling Systems™ Ultra Adhesive (UA) Tape
Ideal for sealing sacks and bags after sampling and dispensing. Sampling Systems™ Ultra Adhesive (UA) Tape is highly versatile and suitable for a range of applications.
Brand:  Sampling Systems™ 8224A-95
 View more versions of this product
Product Code. 15641187
  • ONLINE EXCLUSIVE PRICE
    58.30 EUR
    56.32 EUR / 50m
    Save 1.98 EUR
    3% Off
Only null left
Estimated Shipment: 12-02-2026
Description
  • Uses a special Ultra Adhesive glue, that sticks to practically any surface
  • 50m long roll
  • Easy to cut to required length
  • Individually packed
Specifications
Specifications
Ultra Adhesive Tape
50 x 95 mm
95 mm
White
50 m