missing translation for 'onlineSavingsMsg'
Learn More

Ulixertinib hydrochloride, MedChemExpress

Product Code. 16466848
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16466848 10 mg 10mg
16462917 50 mg 50mg
16409397 5 mg 5mg
16406956 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16466848 Supplier Medchem Express Supplier No. HY15816A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ulixertinib hydrochloride (BVD-523 hydrochloride) is a potent, orally active, highly selective, ATP-competitive and reversible covalent inhibitor of ERK1/2 kinases, with an IC50 of <0.3 nM against ERK2. Ulixertinib hydrochloride inhibits the phosphorylated ERK2 (pERK) and downstream kinase RSK (pRSK) in an A375 melanoma cell line.

Ulixertinib hydrochloride (BVD-523 hydrochloride) is a potent, orally active, highly selective, ATP-competitive and reversible covalent inhibitor of ERK1/2 kinases, with an IC50 of <0.3 nM against ERK2. Ulixertinib hydrochloride inhibits the phosphorylated ERK2 (pERK) and downstream kinase RSK (pRSK) in an A375 melanoma cell line.

Specifications

Chemical Name or Material Ulixertinib hydrochloride
CAS 1956366-10-1
Color White
Purity Grade Notes Research
Molecular Formula C21H23Cl3N4O2
Quantity 10 mg
Synonym BVD-523 hydrochlorideVRT752271 hydrochloride
Solubility Information DMSO : 100 mg/mL (212.86 mM; Need ultrasonic) ∣H2O : 0.1 mg/mL (0.21 mM; ultrasonic and warming and heat to 80°C)
SMILES O=C(C1=CC(C2=CC(NC(C)C)=NC=C2Cl)=CN1)N[C@@H](C3=CC=CC(Cl)=C3)CO.[H]Cl
Molecular Weight (g/mol) 469.79
Formula Weight 469.79
Percent Purity 99.81%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.