missing translation for 'onlineSavingsMsg'
Learn More

Ufenamate, MedChemExpress

Product Code. 16422738
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16422738

Brand: Medchem Express HY100009

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ufenamate (Flufenamic acid butyl ester) is an anthranilic acid-based anti-inflammatory agent. Ufenamate can be used for the research of skin diseases, such as acute and chronic eczema, contact dermatitis, diaper dermatitis, miliaria and atopic dermatitis.

Ufenamate (Flufenamic acid butyl ester) is an anthranilic acid-based anti-inflammatory agent. Ufenamate can be used for the research of skin diseases, such as acute and chronic eczema, contact dermatitis, diaper dermatitis, miliaria and atopic dermatitis.

Specifications

Chemical Name or Material Ufenamate
Physical Form Liquid
CAS 67330-25-0
Color Colorless
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C18H18F3NO2
For Use With (Application) COVID-19-immunoregulation
Synonym Flufenamic acid butyl ester Butyl flufenamate
Solubility Information DMSO : ≥ 44 mg/mL (130.43 mM)
SMILES O=C(OCCCC)C1=CC=CC=C1NC2=CC=CC(C(F)(F)F)=C2
Molecular Weight (g/mol) 337.34
Formula Weight 337.34
Percent Purity 99.85%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.