Learn More
UCMR5 PFAS Mixture 10 μg/mL in Methanol:Water, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| CAS | 39108-34-4, 757124-72-4, 27619-97-2, 763051-92-9, 2991-50-6, 2355-31-9 |
| Quantity | 1 mL |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Methanol/Water |