missing translation for 'onlineSavingsMsg'
Learn More

UBE2D2 Monoclonal Antibody (OTI1A9), TrueMAB™, OriGene

Product Code. 17576713
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17576713 30 μL 30µL
17556713 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17576713 Supplier Origene Technologies Supplier No. TA806527S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

UBE2D2 Monoclonal antibody specifically detects UBE2D2 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen UBE2D2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI1A9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene CLDN3
Gene Accession No. O15551
Gene Alias (E3-independent) E2 ubiquitin-conjugating enzyme D2; 1500034D03Rik; E2 ubiquitin conjugating enzyme UbcH5B; E2 ubiquitin-conjugating enzyme D2; E2(17)KB 2; E2(17)KB2; E217kB; p53-regulated ubiquitin-conjugating enzyme 1; PUBC1; similar to ubiquitin-conjugating enzyme E2D 2; Ubc2e; ubc2e ubiquitin conjugating enzyme; ubc4; UBC4/5; UBC5B; UBCH4; UBCH5B; ube2d2; Ube2d2a; Ubiquitin carrier protein D2; ubiquitin conjugating enzyme 2e; ubiquitin conjugating enzyme E2 D2; ubiquitin conjugating enzyme E2D; ubiquitin conjugating enzyme E2D 2; ubiquitin-conjugating enzyme E2 D2; Ubiquitin-conjugating enzyme E2(17)KB 2; ubiquitin-conjugating enzyme E2-17 kDa 2; ubiquitin-conjugating enzyme E2D 2; ubiquitin-conjugating enzyme E2D 2 (homologous to yeast UBC4/5); ubiquitin-conjugating enzyme E2D 2 (UBC4/5 homolog, yeast); ubiquitin-conjugating enzyme E2D 2A; ubiquitin-protein ligase D2; Unknown (protein for MGC:138112); zgc:55886; zgc:55886 protein; zgc:77149; zmp:0000000618
Host Species Mouse
Immunogen Full length human recombinant protein of human CLDN3 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1365
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.