missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UBD Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5111186
This item is not returnable.
View return policy
Description
Immunogen sequence: NASCLCVHVR SEEWDLMTFD ANPYDSVKKI KEHVRSKTKV PVQDQVLLLG SKILKPRRSL SSYGIDKEKT IHLTLKVVKP Highest antigen sequence indentity to the following orthologs: Mouse - 70%, Rat - 75%.
UBD qualifies as a marker for an interferon response in hepatocellular carcinoma and colon carcinoma but is not significantly overexpressed in cancers lacking a proinflammatory environment. Immunohistochemical studies demonstrated increased UBD expression in HIV-associated nephropathy and in autosomal dominant polycystic kidney disease.
Specifications
| UBD | |
| Polyclonal | |
| Unconjugated | |
| UBD | |
| Diubiquitin; FAT10; GABBR1; Ubd; UBD 3; UBD-3; Ubiquitin D; ubiquitin-like protein FAT10 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10537 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O15205 | |
| UBD | |
| Recombinant Protein corresonding to Human UBD. Recombinant protein control fragment (Product #RP-99571). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction