Description
- Suitable for transparent liquids manual and automatic measurement, foaming liquids, liquid mixture with highly volatile components, and high or low temperature measurements
- The recommended minimum flow through time is 200 seconds
- Calibrated
- With constant for automatic measurements
- Filling quantity of 15 to 20mL
Specifications
Specifications
| Product Type | Viscometer |
| Viscosity Range | 50 to 500 cSt |
| Calibrated | Yes |
| Approximate Constant | 0.5K |
| Certifications/Compliance | DIN 51 562 Part 1, ISO/DIS 3105 (BS-IP-SL) |
| Includes | Ring Marks |
| For Use With (Application) | For Determination of Absolute and Relative Kinematic Viscosity of Liquids With Newtonian Flow Behavior |
| Size | 1.69mm Diameter |
| Volume (Metric) | 15 to 20 mL |