Learn More
SI Analytics™ Ubbelohde Viscometer
Product Code. 12008183 Shop All SI Analytics Products
Click to view available options
Viscosity Range:
0.5 to 3 cSt
0.8 to 5 cSt
1.2 to 10 cSt
10 to 100 cSt
100 to 1000 cSt
1000 to 10,000 cSt
3 to 30 cSt
30 to 300 cSt
5 to 50 cSt
50 to 500 cSt
Approximate Constant:
0.003K
0.005K
0.01K
0.03K
0.05K
0.1K
0.3K
0.5K
10K
1K
Size:
0.47mm Diameter
0.54mm Diameter
0.63mm Diameter
0.84mm Diameter
0.95mm Diameter
1.13mm Diameter
1.50mm Diameter
1.69mm Diameter
2.01mm Diameter
3.60mm Diameter
Unit Size:
Each
Product Code. 12008183
Brand: SI Analytics™ 285400242
Only null left
Add to basket
Add to basket
Determine absolute and relative kinematic viscosity of liquids with Newtonian flow behavior. SI Analytics™ Ubbelohde Viscometer includes ring marks to ensure that they can be checked by means of manual measurements.
  • Suitable for transparent liquids manual and automatic measurement, foaming liquids, liquid mixture with highly volatile components, and high or low temperature measurements
  • The recommended minimum flow through time is 200 seconds
  • Calibrated
  • With constant for automatic measurements
  • Filling quantity of 15 to 20mL
TRUSTED_SUSTAINABILITY
Specifications
Product Type Viscometer
Viscosity Range 10 to 100 cSt
Calibrated Yes
Approximate Constant 0.1K
Certifications/Compliance DIN 51 562 Part 1, ISO/DIS 3105 (BS-IP-SL)
Includes Ring Marks
For Use With (Application) For Determination of Absolute and Relative Kinematic Viscosity of Liquids With Newtonian Flow Behavior
Size 1.13mm Diameter
Volume (Metric) 15 to 20 mL