Learn More
SI Analytics™ Ubbelohde ASTM Viscometer
Product Code. 10099880 Shop All SI Analytics Products
Click to view available options
Approximate Constant:
0.003K
0.005K
0.01K
0.03K
0.3K
Size:
0.36mm Diameter
0.46mm Diameter
0.58mm Diameter
0.78mm Diameter
1.36mm Diameter
Unit Size:
Each
Product Code. 10099880
Brand: SI Analytics™ 285400526
Only null left
Add to basket
Add to basket
Determine absolute and relative kinematic viscosity of liquids with Newtonian flow behavior. SI Analytics™ Ubbelohde ASTM Viscometer includes ring marks to ensure that they can be checked by means of manual measurements.
  • Suitable for transparent liquids manual and automatic measurement, foaming liquids, liquid mixture with highly volatile components, and high or low temperature measurements
  • The recommended minimum flow through time is 200 seconds
  • Calibrated with constant for manual measurements
  • Filling quantity of 15 to 20mL
TRUSTED_SUSTAINABILITY
Specifications
Product Type Viscometer
Calibrated Yes
Approximate Constant 0.005K
ASTM Numbers D 446
Certifications/Compliance ISO/DIS 3105, ASTM D 2515, ASTM D 446
For Use With (Application) Manual Measurements and Automatic Measurements
Size 0.46mm Diameter