missing translation for 'onlineSavingsMsg'
Learn More

Tyrosol Sulfate Sodium Salt, TRC

Product Code. 30454057
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30454057 100 mg 100mg
30452233 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30454057 Supplier Toronto Research Chemicals Supplier No. TRCT947815100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tyrosol Sulfate Sodium Salt
Analyte or Component Names Tyrosol Sulfate Sodium Salt
Shipping Condition Room Temperature
Molecular Formula C8 H9 Na O5 S
Synonym p-Hydroxyphenethyl Alcohol p-(Hydrogen Sulfate) Sodium Salt, 4-(Sulfooxy)benzeneethanol Sodium Salt
InChI Formula InChI=1S/C8H10O5S.Na/c9-6-5-7-1-3-8(4-2-7)13-14(10,11)12;/h1-4,9H,5-6H2,(H,10,11,12);/q;+1/p-1
Solution Type Neat
SMILES OCCc1ccc(OS(=O)(=O)O[Na])cc1
IUPAC Name [4-(2-hydroxyethyl)phenoxy]sulfonyloxysodium
Industry Type Pharmaceutical
Molecular Weight (g/mol) 240.209
Quantity 100 mg
Formula Weight 240.007
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.