Learn More
VACUUBRAND™ Type B Rotary-vane Pump Oil
This vacuum pump oil has excellent viscosity characteristics. Its good chemical resistance, low vapor pressure as well as its better stability when pumping oxidants such as acid and alkaline vapors, makes it superior compared to standard mineral oils. This oil is used for the first filling of RE / RZ / RC series pumps
Brand:  VACUUBRAND™ 20687012
Additional Details : Weight : 18.00000kg
 View more versions of this product
Product Code. 11834431
  • 545.00 EUR / 20 L
Estimated Shipment: 03-07-2024
Description
  • Low vapor pressure even at high temperatures
  • excellent lubricating properties
  • low oil backflow
  • excellent resistance to aging
Specifications
Specifications
Rotary-vane Pump Oil B
<1 x 10-3mbar
94 mm2/sec.
Canister
1 Ea.
20 L