missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TXNDC3 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
178.00 EUR - 383.00 EUR
Specifications
| Antigen | TXNDC3 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18331375
|
Novus Biologicals
NBP3-15525-20UL |
20 μg |
178.00 EUR
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18384174
|
Novus Biologicals
NBP3-15525-100UL |
100 μg |
383.00 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TXNDC3 Polyclonal antibody specifically detects TXNDC3 in Human, Mouse samples. It is validated for Western BlotSpecifications
| TXNDC3 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS, 50% glycerol, pH7.3 | |
| 51314 | |
| IgG | |
| Affinity purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| CILD6Sptrx-2, EC 2.1.1.31, EC 2.5.1.6, NM23-H8, NME8thioredoxin domain-containing protein 3, Spermatid-specific thioredoxin-2, sperm-specific thioredoxin 2, SPTRX2sptrx-2, thioredoxin domain containing 3 (spermatozoa) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 299-588 of human NME8 (NP_057700.3). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title