Learn More
VistaLab™ Technologies 2-Stroke Fixed Volume Ovation BioNatural Pipettes
Product Code. 11956885
Click to view available options
Volume (Metric):
1 μL
5 μL
10 μL
20 μL
25 μL
100 μL
200 μL
250 μL
300 μL
500 μL
Unit Size:
Each
Product Code. 11956885
Brand: VistaLab™ Technologies 12570010
Attain accurate pipetting with this single stroke pipette. Vistalab™ 2-Stroke Fixed Volume Ovation BioNatural Pipettes are excellent for pipetting exact volume of liquids without deviations.
  • Aspirating and Dispensing: A very low-force,form-fitting plunger allows the user to manuallycontrol the speed of aspirating and dispensing with minimal contact or thumb effort.
  • Tip Ejection: A low-force energy release button activates the tip ejector. A gentle push ensures that the forces needed to discard a tip are well below threshold levels recommended by ergonomists
  • Tip Acquisition: The user inserts the nozzle into a tip and lightly presses down until it clicks. No excessive force or jamming is required to ensure a good seal
  • Liquid Blocking Disposable Nozzle: Not every pipetting task requires a filtered tip, but every pipette should be protected from contamination no matter what the task. Ovation comes with a filtered nozzle that does not allow liquid into the body of the device
Specifications
Accuracy ±1.5%
Description 2-stroke
Color Orange
Material Polycarbonate ABS
Model F2
Number of Channels 1
Type Pipettor Ovation
Volume (Metric) 10 μL
Precision 0.005
Quantity 1 ea.
Warranty 2 Year Limited Warranty
Length (English) 9 in.
Width (English) 7 in.
Height (English) 3 in.