Learn More
Lenz Laborglasintrumente Two-Neck Round-Bottom Flasks, Side Neck Parallel
Product Code. 10385921
Click to view available options
Capacity (Metric):
100 mL
500 mL
1 L
2 L
4 L
Standard Taper Joint:
Side Neck NS 14/23
Side Neck NS 29/32
Unit Size:
Each
Product Code. 10385921
Brand: Lenz Laborglasintrumente 03204473
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Distillation flasks
  • With parallel side neck
  • With conical ground joints
  • With inscription label
  • Material: DURAN™
Specifications
Capacity (Metric) 2 L
Material Duran Glass
Standard Taper Joint Side Neck NS 29/32
Diameter (Metric) Outer 166 mm