Learn More
Lenz Laborglasintrumente Two-Neck Round-Bottom Flasks, Side Neck Parallel
Click to view available options
Capacity (Metric):
100 mL
500 mL
1 L
2 L
4 L
Standard Taper Joint:
Side Neck NS 14/23
Side Neck NS 29/32
Unit Size:
Each
Description
- Distillation flasks
- With parallel side neck
- With conical ground joints
- With inscription label
- Material: DURAN™
Specifications
Specifications
| Capacity (Metric) | 1 L |
| Material | Duran Glass |
| Standard Taper Joint | Side Neck NS 29/32 |
| Diameter (Metric) Outer | 131 mm |