Learn More
Lenz Laborglasinstrumente™ Two-Neck Round-Bottom Flasks, Side Neck Angled
Product Code. p-7222493
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1 L
2 L
Standard Taper Joint:
Side Neck 29/32
Side Neck NS 14/23
Side Neck NS 19/26
Unit Size:
Each
Product Code. 10519951
Brand: Lenz Laborglasinstrumente™ 03214173
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Distillation flasks acc. to DIN 12394 and in addition to DIN standard
  • With NS
  • With angled side neck (20°)
  • With inscription label
  • Material: DURAN™
Specifications
Capacity (Metric) 2 L
Material Duran Glass
Standard Taper Joint Side Neck NS 14/23