Learn More
Labplas Twirl'EM™ Large Format Sampling Bags with Tabs
Click to view available options
Capacity (Metric):
6.75 L
Unit Size:
Pack of 250
Description
Polyethylene sampling bags with large openings
- Large sized
- Contaminant-free
- Virgin polyethylene
- Large bag mouth opening
- Easy to use pull tabs
Specifications
Specifications
| Description | Regular Tabs |
| Capacity (Metric) | 6.75 L |
| Closure Type | 2 Flat Wires with Tabs |
| Material | Polyethylene |
| Sterility | Sterile |
| Thickness (English) | 4 mil |
| Dimensions (L x W) | 18 x 12 in. (457 x 305mm) |
| Length (English) | 18 in. |
| Length (Metric) | 457 mm |
| Width (English) | 12 in. |
| Show More |