missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ GlobalMark™ Squeegee
Description
- For Use With: Global Mark Printers
Specifications
Specifications
| Product Type | Squeegee |
| Color | Blue |
| For Use With (Equipment) | BBP30 Label Printer, BBP31 Sign and Label Printer, BBP35 Multi-Color Sign and Label Printer, BBP37 Color and Cut Sign and Label Printer |
| Format | Squeegee |
| Quantity | 1 Ea. |
| Alternate Part Number | Squeegee |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?