Learn More
RETSCH Tungsten Carbide Grinding Balls
Product Code. 10166641 Shop All Retsch Products
Change view
Click to view available options
For Use With (Equipment):
Planetary Ball Mill PM 400
Reaction Vials
Diameter (Metric):
4 mm
5 mm
20 mm
30 mm
40 mm
Unit Size:
Each
Pack of 200
Product Code. For Use With (Equipment) Diameter (Metric) unitSize
10166641 Reaction Vials 4 mm Pack of 200
10166451 Planetary Ball Mill PM 400 40 mm Each
10509331 Planetary Ball Mill PM 400 30 mm Each
10298510 Planetary Ball Mill PM 400 20 mm Each
10156641 Reaction Vials 5 mm Pack of 200
5 options
Product Code. 10166641
Brand: RETSCH 22.455.0005
Only null left
Specifications
Product Type Grinding Balls
Diameter (Metric) 4 mm
For Use With (Equipment) Reaction Vials
Material Tungsten Carbide