Learn More
Honeywell™ Tuff Cut Nit Gloves
Description
- Knitted glove in Kevlar™, high tenacity polyamide, steel fiber
- The knitting technique helps to obtain a glove that conforms snugly to the hand and good dexterity
- The glove offers maximum comfort and added sturdiness
- The Kevlar™ fibrerensures good comfort in contact with the hand
- The use of foam nitrile makes the glove more flexible than standard nitrile
- The composition of the glove offers outstanding cutting resistance (level 5)
- The nitrile foam coating brings good resistance and good grip to oil and grease
- The 3/4 coating increase the protection to oil
- Color make it more soilproof, thus ensuring a longer last
Specifications
Specifications
| Certifications/Compliance | EN 420-2003 EEC Basic standards, EN 388-2016 Mechanical Hazards, EN407-2004 : Thermal risks |
| Description | 10 gauge |
| Coating Material | Nitrile Foam |
| Color | Black, Yellow |
| Material | Polyamide, Stainless Steel Fiber, Dupont™ Kevlar™ Aramid Fibers |
| Size | 9 |
| Length (Metric) | 30 cm |