Learn More
Honeywell™ Tuff Cut Nit Gloves
Product Code. 15795878
Change view
Click to view available options
:
Size: 11
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15795878 Size: 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15795878 Supplier Honeywell™ Supplier No. 223253311
Only null left
Add to basket
Add to basket
Honeywell™ Tuff Cut Nit Gloves for handling heavy parts with high cut risks in an oily or greasy environment.
  • Knitted glove in Kevlar™, high tenacity polyamide, steel fiber
  • The knitting technique helps to obtain a glove that conforms snugly to the hand and good dexterity
  • The glove offers maximum comfort and added sturdiness
  • The Kevlar™ fibrerensures good comfort in contact with the hand
  • The use of foam nitrile makes the glove more flexible than standard nitrile
  • The composition of the glove offers outstanding cutting resistance (level 5)
  • The nitrile foam coating brings good resistance and good grip to oil and grease
  • The 3/4 coating increase the protection to oil
  • Color make it more soilproof, thus ensuring a longer last
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 420-2003 EEC Basic standards, EN 388-2016 Mechanical Hazards, EN407-2004 : Thermal risks
Description 10 gauge
Coating Material Nitrile Foam
Color Black, Yellow
Material Polyamide, Stainless Steel Fiber, Dupont™ Kevlar™ Aramid Fibers
Size 9
Length (Metric) 30 cm