Learn More
Upchurch Scientific Tubing sleeve PEEK(TM) blue 13 inch (33mm) 1/16inch O.D., 250µm I.D. for 225 to 265µm O.D. tubing
Brand:  Upchurch Scientific 02017-89
Tubing sleeve PEEK(TM) blue 13 inch (33mm) 1/16inch O.D., 250µm I.D. for 225 to 265µm O.D. tubing
Product Code. 13033090
  • 13.20 EUR / Each
Only null left
Estimated Shipment: 28-01-2026