Learn More
Upchurch Scientific Tubing sleeve PEEK(TM) blue 13 inch (33mm) 1/16inch O.D., 250µm I.D. for 225 to 265µm O.D. tubing
Brand: Upchurch Scientific 02017-89
Tubing sleeve PEEK(TM) blue 13 inch (33mm) 1/16inch O.D., 250µm I.D. for 225 to 265µm O.D. tubing