Learn More
Gilson™ Tubing for Bottle-Hand Operator
Shop All Gilson Products
Click to view available options
Overall Length (Metric):
1800 mm
Unit Size:
Each
Description
- Provided in lengths of 1800mm
- For use with Safe Aspiration Stations
- Supplied individually
Specifications
Specifications
| Description | For use with Safe Aspiration Stations |
| Overall Length (Metric) | 1800 mm |
| Packaging Quantity | 1 |
| Tubing Size | 1800mm |