Learn More
Masterflex™ Tubing adapter NPT (F) PVDF
Product Code. 12418680
Product Code. 12418680 Supplier Masterflex™ Supplier No. 3120111
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Meet USP Class VI for pharmaceutical use and FDA, USDA, and 3A standards for food contact
  • All fittings are compatible with Tri-Clamp™ connections
  • Excellent chemical resistance; low leachable characteristics
Specifications
Description Tubing Adapter; , 3/4 in. mini to 1/4 in. NPT (F)
For Use With Masterflex L/S and I/P sanitary tubing connection
Outlet Port Sanitary x NPT(F)
Thread Size 1/4 in.
Includes Connection Type: 3/4 in. mini to 1/4 in. NPT(F)
Material (Seal) PVDF
Product Type Vacuum Tubing