Description
The Tuberin/TSC2 antibody from Proteintech is a mouse monoclonal antibody generated with peptide of human Tuberin/TSC2. This antibody recognizes human, mouse, rat antigen. The Tuberin/TSC2 antibody has been validated for the following applications: WB, IHC, IF/ICC, ELISA analysis.
Specifications
Specifications
| Antigen | Tuberin/TSC2 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA |
| Classification | Monoclonal |
| Clone | 1A10B2 |
| Concentration | 1000 μg/mL |
| Conjugate | Unconjugated |
| Gene Symbols | FLJ43106; LAM; TSC4 |
| Host Species | Mouse |
| Immunogen | Peptide |
| Purification Method | Protein A purification |
| Show More |