Learn More
BOLA Tube End Fittings
Product Code. 10787692
Change view
Click to view available options
Color:
Black
Blue
Green
Orange
Red
Violet
White
Yellow
Diameter (Metric) Outer:
1.6 mm
1.61 mm
3.2 mm
Unit Size:
Pack of 10
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Diameter (Metric) Outer unitSize
10787692 Yellow 1.6 mm Pack of 10
10593822 White 1.61 mm Pack of 10
10165161 White 3.2 mm Pack of 10
10506132 White 1.6 mm Pack of 10
10308951 Black 3.2 mm Pack of 10
10174491 Red 1.6 mm Pack of 10
10083461 Red 3.2 mm Pack of 10
10093461 Orange 3.2 mm Pack of 10
10369041 Yellow 3.2 mm Pack of 10
10698831 Green 1.6 mm Pack of 10
10707532 Blue 1.6 mm Pack of 10
10319191 Violet 1.6 mm Pack of 10
10175451 Violet 3.2 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 10787692 Supplier BOLA Supplier No. F70230
Only null left
Add to basket
Add to basket
Tube End Fittings with Washers
  • Tube end fittings.
  • With male thread UNF ¼ in. 28 G washers made of PA are included in delivery.
  • Packing unit: 10 pieces, differing ordering quantities are rounded up to factor 10.
Specifications
Color Yellow
Diameter (Metric) Outer 1.6 mm
Diameter (Metric) Inner 0.8 mm