Learn More
Ismatec™ Tube Connector, Straight, Steel 18/8
Product Code. 11996036
Change view
Click to view available options
Diameter (Metric) Inner:
For tubing with I.D. 0.58 mm
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner unitSize
11996036 For tubing with I.D. 0.58 mm Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11996036 Supplier Ismatec™ Supplier No. ISM581
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Standard tube connector for pump tubing.
  • Standard connector, straight
  • Suitable for use with pump tubing
  • Steel (18/8) construction
Specifications
Diameter (Metric) Inner For tubing with I.D. 0.58 mm
Diameter (Metric) Outer For tubing with I.D. 0.90 mm
Overall Length (Metric) 15 mm
For Use With Pump tubing
Packaging Quantity 6/Pk
Shape Straight