Learn More
SCAT™ Tube Connector, Straight
Product Code. 10631021 Shop All SCAT Europe Products
Click to view available options
Diameter (Metric):
5 to 7 mm
6 to 8 mm
6.2 to 7.5 mm
9.5 to 10 mm
Unit Size:
Each
Product Code. 10631021
Brand: SCAT™ 117816
Only null left
Add to basket
Add to basket
Safe connection of tubes of different sizes: directly and without technical aids. For safe collection of liquid HPLC waste, and many other applications in the laboratory!
SCAT™ connectors are available in the following versions:
  • Conical connectors
  • Y-connectors
  • Tube connectors
  • Capillary connectors
Specifications
Diameter (Metric) 6 to 8 mm
Autoclavable Autoclavable
Material Polypropylene
For Use With Tubing and Capillaries
For Use With (Application) HPLC/UHPLC Systems
Color Transparent
Product Type Tubing Connector