Learn More
TTYH3 Rabbit, anti-Human, Polyclonal, Proteintech
Product Code. 18048945 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18048945 150 μL 150µL
18079345 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18048945 Supplier Proteintech Supplier No. 247071AP150UL
Rabbit Polyclonal Antibody
The TTYH3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human TTYH3. This antibody recognizes human antigen. The TTYH3 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Antigen TTYH3
Applications Western Blot, ELISA
Concentration 650 μg/mL
Gene Symbols hTTY3, KIAA1691, Protein tweety homolog 3, TTYH3, tweety homolog 3 (Drosophila)
Host Species Rabbit
Quantity 150 μL
Primary or Secondary Primary
Gene ID (Entrez) 80727
Target Species Human
Content And Storage Store at -20°C. Stable for one year after shipment.
Antibody Molecular Weight 150 kDa
Form Liquid
Isotype IgG