Learn More
TTNPB, MedChemExpress
Product Code. 16476738
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16476738 10 mg 10mg
16499287 5 mg 5mg
16472817 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16476738 Supplier Medchem Express Supplier No. HY15682
MedChemExpress TTNPB is a highly potent RAR agonist. Competitive binding assays using human RARs yield IC50s of α=5.1 nM, β= 4.5 nM, and γ=9.3 nM, respectively.
TTNPB is a highly potent RAR agonist. Competitive binding assays using human RARs yield IC50s of α=5.1 nM, β= 4.5 nM, and γ=9.3 nM, respectively.
Specifications
Chemical Name or Material TTNPB
CAS 71441-28-6
Color White
Purity Grade Notes Research
Molecular Formula C24H28O2
Quantity 10 mg
Synonym Ro 13-7410 Arotinoid acid AGN191183
Solubility Information DMSO : 7.58 mg/mL (21.75 mM; Need ultrasonic)
SMILES O=C(O)C1=CC=C(/C=C(C2=CC=C3C(C)(C)CCC(C)(C)C3=C2)\C)C=C1
Molecular Weight (g/mol) 348.48
Formula Weight 348.48
Percent Purity 98.02%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death