missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TSPAN6 Polyclonal Antibody

Product Code. 17148453
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17148453 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17148453 Supplier Invitrogen™ Supplier No. PA5110528

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: NSFKNNYEKA LKQYNSTGDY RSHAVDKIQN TLHCCGVTDY RDWTDTNYYS EKGFPKSCCK LEDCTPQRDA DKVNNEGCFI K.

The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. This encoded protein is a cell surface glycoprotein and is highly similar in sequence to the transmembrane 4 superfamily member 2. The use of alternate polyadenylation sites has been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSPAN6
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.85 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene TSPAN6
Gene Accession No. O70401
Gene Alias 6720473L21Rik; A15 homolog; AI316786; Putative NF-kappa-B-activating protein 321; T245; T245 protein; tetraspan TM4SF; tetraspanin 6; tetraspanin TM4-D; Tetraspanin-6; Tm4sf; TM4SF6; Transmembrane 4 superfamily member 6; TSPAN6; Tspan-6; UNQ767/PRO1560
Gene Symbols TSPAN6
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 120-200 of human TSPAN6 (NP_003261.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56496
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.