missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TSPAN3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA528514
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, HeLa, mouse kidney. Predicted reactivity: Mouse (84%), Rat (83%), Pig (84%), Rhesus Monkey (86%), Bovine (85%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. The use of alternate polyadenylation sites has been found for this gene. Two alternative transcripts encoding different isoforms have been described.
Specifications
| TSPAN3 | |
| Polyclonal | |
| Unconjugated | |
| TSPAN3 | |
| 1700055K04Rik; H41D4 protein; OAP-1; OSP-associated protein 1; tetraspan 3; tetraspan TM4SF; tetraspanin 3; tetraspanin TM4-A; tetraspanin TM4-A homolog; tetraspanin-3; TM4-A; Tm4sf8; Transmembrane 4 superfamily member 8; TSPAN3; TSPAN-3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10099, 56434 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| O60637, Q9QY33 | |
| TSPAN3 | |
| Recombinant protein encompassing a sequence within the center region of human TSPAN3. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction