missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSGA2 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-20992
This item is not returnable.
View return policy
Description
TSGA2 Polyclonal specifically detects TSGA2 in Mouse samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| TSGA2 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| Cancer/testis antigen 79, CT79, FLJ32753, h-meichroacidin, Male meiotic metaphase chromosome-associated acidic protein, Meichroacidin, radial spoke head 1 homolog, radial spoke head 1 homolog (Chlamydomonas), RSP44, RSPH10A, testes specific A2 homolog, testes specific gene A2 homolog, testis specific A2 homolog, testis specific A2 homolog (mouse), Testis-specific gene A2 protein, TSA2MGC126568, TSGA2MGC141927 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Mouse reactivity determined by Western Blot. Expected from sequence similarity: Rat, Cow. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.1 - 0.3 ug/ml, Peptide ELISA Detection limit 1:4000 | |
| NP_543136.1 | |
| RSPH1 | |
| Peptide with sequence C-HLNHRYQGKFLNKNP corresponding to internal region according to NP_543136.1. | |
| 0.1 mg | |
| Cell Cycle and Replication | |
| 89765 | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction