Learn More
Thermo Scientific™ Tryptone Soya Agar with Disinhibitor
Description
Effortlessly sample surfaces for environmental monitoring after cleaning and disinfection using Thermo Scientific™ Tryptone Soy Agar with Disinhibitor containing lecithin, polysorbate 80, sodium thiosulfate and Tween 80 to neutralize surface disinfectants and aid in microorganism recovery.
Use Tryptone Soya Agar with Disinhibitor for sterility control while neutralizing the possible remains of disinfectants and preservatives.
- Aids in recovery of microorganisms while neutralizing disinfectants and preservatives.
- Conforms to formulations detailed in International Pharmacopoeia.
- Ready-to-use convenience of prepared media.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
| Color | Ivory, transparent |
| Content And Storage | 6°C to 12°C |
| Description | Tryptone Soya Agar with Disinhibitor |
| Format | Poured Plate |
| Manufacturing Origin | Germany |
| Product Type | Agar |
| Fill Weight | 17g ± 5 % |
| Packaging Type | Flow Wrapped - Single Layer |
| Quantity | 10/Pk. |