missing translation for 'onlineSavingsMsg'
Learn More

Trovirdine, MedChemExpress

Product Code. 16459057
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16459057

Brand: Medchem Express HY15349

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Trovirdine inhibits HIV-1 RT with an IC50 of 7 nM when employing heteropolymeric primer/template (oligo-DNA/ribosomal RNA)and dGTP as substrate.

Trovirdine inhibits HIV-1 RT with an IC50 of 7 nM when employing heteropolymeric primer/template (oligo-DNA/ribosomal RNA)and dGTP as substrate.

Specifications

Chemical Name or Material Trovirdine
CAS 149488-17-5
Color White
Purity Grade Notes Research
Molecular Formula C13H13BrN4S
Quantity 5 mg
Synonym LY300046
Solubility Information DMSO : 100 mg/mL (296.52 mM; Need ultrasonic)
SMILES S=C(NCCC1=NC=CC=C1)NC2=NC=C(Br)C=C2
Molecular Weight (g/mol) 337.24
Formula Weight 337.24
Percent Purity 98.59%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.