Learn More
Thermo Scientific™ Trolley for VersaCool™ Refrigerated Circulating Bath
Product Code. 15409539 Shop All Thermo Scientific Products
Click to view available options
Type:
Trolley
Unit Size:
Each
Product Code. 15409539
Brand: Thermo Scientific™ 230000004
Only null left
Add to basket
Add to basket
Easily move your VersaCool between fume hood, benches and various applications with the trolley.
Easily move your Thermo Scientific™ VersaCool™ between fume hood, benches and various applications with our Trolley. The castors provide a stable base to ensure your unit is secured.
Certification/Compliance: UL Listed
TRUSTED_SUSTAINABILITY
Specifications
Description Trolley for VersaCool
For Use With (Equipment) Thermo Scientific VersaCool Refrigerated Circulating Bath
Type Trolley