missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRMU Monoclonal Antibody (OTI1C6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505698S
This item is not returnable.
View return policy
Description
TRMU Monoclonal antibody specifically detects TRMU in Human samples. It is validated for Western BlotSpecifications
| TRMU | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O75648 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 55687 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Western Blot | |
| OTI1C6 | |
| Unconjugated | |
| TRMU | |
| 1110005N20Rik; 1600025P05Rik; AI314320; LCAL3; lung cancer associated lncRNA 3; mitochondrial 5-methylaminomethyl-2-thiouridylate-methyltransferase; mitochondrial tRNA-specific 2-thiouridylase 1; MTO2; MTO2 homolog; MTU1; TRMT; Trmt1; Trmu; tRNA (5-methylaminomethyl-2-thiouridylate)-methyltransferase 1; tRNA 5-methylaminomethyl-2-thiouridylate methyltransferase; tRNA mitochondrial 2-thiouridylase; TRNT1 | |
| Full length human recombinant protein of human TRMU produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction