missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRMT11 Polyclonal Antibody
Description
Immunogen sequence: FSVLEDYGLD PNCIPENPHN IYFGRWIADG QRELIESYSV KKRHFIGNTS MDAGLSFIMA NHGKVKENDI VFDPFVGTGG.
TRMT11 belongs to the methyltransferase superfamily. It is a catalytic subunit of an S-adenosyl-L-methionine-dependent tRNA methyltransferase complex that mediates the methylation of the guanosine nucleotide at position 10 (m2G10) in tRNAs.
Specifications
Specifications
| Antigen | TRMT11 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | TRMT11 |
| Gene Accession No. | Q7Z4G4 |
| Gene Alias | 2410075D05Rik; 3110045I18Rik; AW213713; C6orf75; MDS024; putative RNA methylase MDS024; TRM11; Trmt11; TRMT11-1; tRNA (guanine(10)-N2)-methyltransferase homolog; tRNA guanosine-2'-O-methyltransferase TRM11 homolog; tRNA guanosine-2'-O-methyltransferase TRM11-like protein-like protein; tRNA methyltransferase 11; tRNA methyltransferase 11 homolog; tRNA methyltransferase 11 homolog (S. cerevisiae) |
| Gene Symbols | TRMT11 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?