Learn More
[Tris(trimethylsilyl)]phosphine, 98%, Thermo Scientific Chemicals
Product Code. p-7203428 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 mL
5 mL
Unit Size:
1mL
5mL
Product Code. 10547231
Brand: Thermo Scientific Acros 291700010
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 15573-38-3
Molecular Formula C9H27PSi3
MDL Number MFCD00015487
InChI Key OUMZKMRZMVDEOF-UHFFFAOYSA-N
Synonym tris trimethylsilyl phosphine, tris trimethylsilyl phosphane, phosphine, tris trimethylsilyl, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl disilaphosphane, tris trimethylsilyl phosphine in hexane, pubchem6456, acmc-1btki, tris-trimethylsilyl phosphine, phosphine,tris trimethylsilyl
PubChem CID 272683
IUPAC Name tris(trimethylsilyl)phosphane
SMILES C[Si](C)(C)P([Si](C)(C)C)[Si](C)(C)C
Specifications
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.86g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle packed in tin can
Linear Formula [(CH3)3Si]3P
Quantity 1 mL
Molecular Weight (g/mol) 250.544
Formula Weight 250.54
Physical Form Liquid
Chemical Name or Material [Tris(trimethylsilyl)]phosphine, 98%