missing translation for 'onlineSavingsMsg'
Learn More

Tris(methylcyclopentadienyl)yttrium(III), 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11304487
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11304487 1 g 1g
11394477 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11304487 Supplier Thermo Scientific Alfa Aesar Supplier No. 039576.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 329735-72-0
Molecular Formula C18H21Y
Molecular Weight (g/mol) 326.272
MDL Number MFCD00145485
InChI Key OXKRLQPVIZJLDT-UHFFFAOYSA-N
Synonym tris methylcyclopentadienyl yttrium iii, yttrium tris 2-methylcyclopenta-1,3-dien-1-ide, yttrium 3+ ion tris 2-methylcyclopenta-1,3-dien-1-ide
PubChem CID 73991964
IUPAC Name 2-methylcyclopenta-1,3-diene;yttrium(3+)
SMILES CC1=[C-]CC=C1.CC1=[C-]CC=C1.CC1=[C-]CC=C1.[Y+3]

Specifications

Color Yellow
Packaging Ampule
Quantity 1 g
UN Number UN3181
Sensitivity Air and moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 326.27
Percent Purity 99.9%
Physical Form Crystalline
Assay (REO)
Chemical Name or Material Tris(methylcyclopentadienyl)yttrium(III)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.