missing translation for 'onlineSavingsMsg'
Learn More

Tris[[2-(tert-butoxycarbonyl)ethoxy]methyl]methylamine, Tris[[2-(tert-butoxycarbonyl)ethoxy]methyl]methylamine, TRC

Product Code. 17984315
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17984315 250 mg 250mg
17952065 5 g 5g
17951625 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17984315 Supplier Toronto Research Chemicals Supplier No. TRCT876200250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C25 H47 N O9
Molecular Weight (g/mol) 505.642
Synonym 3,3'-[[2-Amino-2-[[3-(1,1-dimethylethoxy)-3-oxopropoxy]methyl]-1,3-propanediyl]bis(oxy)]bispropanoic Acid 1,1'-Bis(1,1-dimethylethyl) Ester, 3,3'-[[2-Amino-2-[[3-(1,1-dimethylethoxy)-3-oxopropoxy]methyl]-1,3-propanediyl]bis(oxy)]bispropanoic Acid Bis(1,1-dimethylethyl) Ester
IUPAC Name tert-butyl 3-[2-amino-3-[3-[(2-methylpropan-2-yl)oxy]-3-oxopropoxy]-2-[[3-[(2-methylpropan-2-yl)oxy]-3-oxopropoxy]methyl]propoxy]propanoate
SMILES CC(C)(C)OC(=O)CCOCC(N)(COCCC(=O)OC(C)(C)C)COCCC(=O)OC(C)(C)C

Specifications

CAS 175724-30-8
Quantity 250 mg
InChI Formula InChI=1S/C25H47NO9/c1-22(2,3)33-19(27)10-13-30-16-25(26,17-31-14-11-20(28)34-23(4,5)6)18-32-15-12-21(29)35-24(7,8)9/h10-18,26H2,1-9H3
Formula Weight 505.3251
Chemical Name or Material Tris[(2-(tert-butoxycarbonyl)ethoxy)methyl]methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.